missing translation for 'onlineSavingsMsg'
Learn More

CD38 Antibody (240726) [DyLight 650], Novus Biologicals™

Product Code. 30733143 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733143 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733143 Supplier Novus Biologicals Supplier No. FAB24041W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD38
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 240726
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias 2'-phospho-ADP-ribosyl cyclase, 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase (EC:2.4.99.20), 2'-phospho-cyclic-ADP-ribose transferase, ADPRC 1, ADPRC1, ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1, cADPr hydrolase 1, CD_antigen: CD38, CD38, CD38 antigen, CD38 antigen (p45), CD38 molecule, cluster of differentiation 38, Cyclic ADP-ribose hydrolase 1, EC 2.4.99.20, EC 3.2.2.6, EC:3.2.2.6, NAD(+) nucleosidase, T10
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CD38, Met1-Ile300, Accession # P28907
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, B Cell Development and Differentiation Markers, Cancer, Cell Biology, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 952
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.