missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD37 Monoclonal Antibody (IPO-24)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA17627
This item is not returnable.
View return policy
Description
MA1-7627 detects CD37 from human samples. MA1-7627 has been successfully used in ELISA, flow cytometry, immunoprecipitation and immunohistochemistry procedures. The MA1-7627 immunogen is a A B cell line.
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. It may play a role in T-cell-B-cell interactions. Alternate splicing results in multiple transcript variants encoding different isoforms.
Specifications
| CD37 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| P11049 | |
| CD37 | |
| A B cell line. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation | |
| IPO-24 | |
| Unconjugated | |
| CD37 | |
| CD37; CD37 antigen; CD37 molecule; cell differentiation antigen 37; GP52-40; Leukocyte antigen CD37; leukocyte surface antigen CD37; tetraspanin-26; Tspan26; Tspan-26 | |
| Mouse | |
| Protein A | |
| RUO | |
| 951 | |
| 4°C short term, -80°C long term | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction