missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD37 Monoclonal Antibody (2B8)
Description
MA5-15492 targets CD37 in indirect ELISA, IF applications and shows reactivity with Human and mouse samples. The MA5-15492 immunogen is purified recombinant fragment of CD37 expressed in E. Coli.. MA5-15492 detects CD37 which has a predicted molecular weight of approximately 31.7kDa.
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. It may play a role in T-cell-B-cell interactions. Alternate splicing results in multiple transcript variants encoding different isoforms.
Specifications
Specifications
| Antigen | CD37 |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2B8 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene | CD37 |
| Gene Accession No. | P11049, Q61470 |
| Gene Alias | CD37; CD37 antigen; CD37 molecule; cell differentiation antigen 37; GP52-40; Leukocyte antigen CD37; leukocyte surface antigen CD37; tetraspanin-26; Tspan26; Tspan-26 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?