missing translation for 'onlineSavingsMsg'
Learn More

CD37 Antibody (MB-1), DyLight 594, Novus Biologicals™

Product Code. 18395573
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395573 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18395573 Supplier Bio-Techne Supplier No. NBP311968DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD37 Monoclonal antibody specifically detects CD37 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD37
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MB-1
Conjugate DyLight 594
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CD37 antigentetraspanin-26, CD37 molecule, cell differentiation antigen 37, GP52-40, leukocyte antigen CD37, MGC120234, Tetraspanin-26, Tspan-26, TSPAN26leukocyte surface antigen CD37
Host Species Rabbit
Immunogen BALB/c mice were immunized with lymph node cells from a patient with nodular poorly differentiated lymphocytic lymphoma. Immunophenotyping studies of these cells identified them as B cells expressing SIgG/lambda, as well as HLA-DR. Leu-12, and B1.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 951
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.