missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD37 Antibody (IPO-24 (ZC37-24)), Alexa Fluor™ 405, Novus Biologicals™
Description
CD37 Monoclonal antibody specifically detects CD37 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Specifications
Specifications
| Antigen | CD37 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | IPO-24 (ZC37-24) |
| Conjugate | Alexa Fluor 405 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD37 antigentetraspanin-26, CD37 molecule, cell differentiation antigen 37, GP52-40, leukocyte antigen CD37, MGC120234, Tetraspanin-26, Tspan-26, TSPAN26leukocyte surface antigen CD37 |
| Host Species | Rabbit |
| Immunogen | This antibody was raised by immunizing mice with spleen cells of a patient with hairy cell leukemia. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?