missing translation for 'onlineSavingsMsg'
Learn More

CD37 Antibody (424925), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals FAB4625MFV610

407.15 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


Product Code. 18557497

  • £510.00 / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD37 Monoclonal antibody specifically detects CD37 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD37
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Mouse
0.1 mL
Cellular Markers, Immunology
951
Store at 4C in the dark.
Flow Cytometry, CyTOF
424925
Flow Cytometry, CyTOF-ready
CD37 antigentetraspanin-26, CD37 molecule, cell differentiation antigen 37, GP52-40, leukocyte antigen CD37, MGC120234, Tetraspanin-26, Tspan-26, TSPAN26leukocyte surface antigen CD37
Mouse myeloma cell line NS0-derived recombinant human CD37, Accession # Q96LM7
RUO
Primary
Human
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.