missing translation for 'onlineSavingsMsg'
Learn More

CD366 (TIM-3) Mouse anti-Mouse, PE, Clone: 5D12/TIM-3 (also known as 5D12), BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 566346

Product Code. 15844969

  • ONLINE EXCLUSIVE PRICE
    £255.00
    £244.82 / Each
    Save £10.18
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 5D12 monoclonal antibody specifically binds to NKp80 which is also known as C-type lectin domain family 5 member C (CLEC5C). NKp80 is a single-pass type II transmembrane glycoprotein that contains a C-type lectin extracellular domain and is expressed as a ñ80 kDa disulfide-linked homodimer. NKp80 is encoded by KLRF1 /(Killer cell lectin-like receptor subfamily F member 1) which maps to the NK-gene complex (NKC). NKp80 is expressed on most NK cells, and subsets ofγδ T cellsand effector-memory CD8αβT cells. NKp80 is not present in rodents. NKp80 is an activating coreceptor that induces NK cell-mediated cytotoxicity and proinflammatory cytokine (eg, IFN-γand TNF) production upon interaction with its ligand, AICL (Activation induced C type lectin). AICL is encoded by CLEC2B /that is selectively expressed on myeloid cells including activated monocytes, macrophages, and granulocytes. NKp80 can reportedly transduce signals through an atypical hemi-ITAM-like sequence motif and Syk kinase to trigger cellular responses.

R-phycoerythrin (PE) is an accessory photosynthetic pigment found in red algae. It exists in vitro as a 240-kDa protein with 23 phycoerythrobilin chromophores per molecule. This makes PE one of the brightest fluorochromes for flow cytometry applications, but its photobleaching properties make it unsuitable for fluorescence microscopy.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD366 (TIM-3)
5D12/TIM-3 (also known as 5D12)
PE
Cd366, T-cell membrane protein, Tim3, TIMD-3, Timd3, HAVcr-2, Havcr2,
Mouse TIM-3 Recombinant Protein
100 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
Affinity Purified
RUO
Mouse
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.