Learn More
CD360 (IL-21 Receptor) Monoclonal Antibody (2SX21R), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£116.00 - £277.00
Specifications
Antigen | CD360 (IL-21 Receptor) |
---|---|
Clone | 2SX21R |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15559246
|
Affymetrix eBioscience
46-3601-42 |
100 Tests |
£277.00
100 tests |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15549246
|
Affymetrix eBioscience
46-3601-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This 2SX21R monoclonal antibody reacts with human CD360 (Interleukin-21 Receptor, IL-21R). IL-21R forms a heterodimeric receptor complex with the common gamma chain to form a functional receptor for IL-21. IL-21 is a class I cytokine of the common gamma chain family of cytokines and signals through the IL-21R complex to activate the JAK/STAT (Janus Kinase/Signal Transducers and Activators of Transcription) signaling pathway. Activation of the JAK/STAT pathway by IL-21 promotes transcription of genes responsive to STAT1 and STAT3. IL-21 is mainly produced by CD4+ T cells and mediates the proliferation, differentiation, and effector functions of cells expressing IL-21R such as B cells, T cells, NK cells, dendritic cells and other populations of myeloid cells. IL-21 and its receptor are implicated in the pathogenesis of chronic viral infections, autoimmunity, allergy, organ transplantation, and cancer. Applications Reported: This 2SX21R antibody has been reported for use in flow cytometric analysis. Applications Tested: This 2SX21R antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
CD360 (IL-21 Receptor) Monoclonal antibody specifically detects CD360 (IL-21 Receptor) in Human samples. It is validated for Flow Cytometry.Specifications
CD360 (IL-21 Receptor) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q9HBE5 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
2SX21R | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Human | |
50615 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.