missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD36/SR-B3 Antibody (324205), DyLight 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB25191M
This item is not returnable.
View return policy
Description
CD36/SR-B3 Monoclonal specifically detects CD36/SR-B3 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
| CD36/SR-B3 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 948 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry | |
| 324205 | |
| Flow Cytometry, Immunohistochemistry | |
| BDPLT10, CD_antigen: CD36, CD36, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor), CD36 molecule (thrombospondin receptor), CHDS7, cluster determinant 36, FAT, Fatty acid translocase, Glycoprotein IIIb, GP3B, GP4, GPIIIB, GPIV, PAS IV, PAS-4, PAS-4 protein, PASIV, Platelet collagen receptor, platelet glycoprotein 4, SCARB3, scavenger receptor class B, member 3, Thrombospondin receptor | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse CD36/SR-B3, Gly30-Lys439, Accession # Q08857 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction