missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD36 Recombinant Rabbit Monoclonal Antibody (23GB3370), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | CD36 |
|---|---|
| Clone | 23GB3370 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088184
|
Invitrogen™
MA553374 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089180
|
Invitrogen™
MA553375 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD36 Recombinant Monoclonal antibody specifically detects CD36 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| CD36 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P16671, Q07969, Q08857 | |
| 12491, 29184, 948 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3370 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| adipocyte membrane protein; BDPLT10; Cd36; CD36 antigen; CD36 antigen (collagen type I receptor, thrombospondin receptor); CD36 molecule; CD36 molecule (thrombospondin receptor); CD36 protein; CHDS7; cluster determinant 36; collagen type I receptor thrombospondin receptor; collagen type I receptor, thrombospondin receptor; FAT; FAT/CD36; fatty acid translocase; fatty acid translocase/CD36; fatty acid transport protein; glycoprotein IIIb; GP3B; GP4; GPIIIB; GPIV; I79_011940; Leukocyte differentiation antigen CD36; PAS IV; PAS4; PAS-4; PAS-4 protein; PASIV; Platelet collagen receptor; Platelet glycoprotein 4; platelet glycoprotein IV; Scarb3; scavenger receptor class B, member 3; SR-B3; Thrombospondin receptor | |
| A synthetic peptide of human CD36 (350-400AA). | |
| Primary | |
| -20°C | |
| CD36 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title