missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD36 Rabbit anti-Hamster, Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18836-1-AP-20UL
This item is not returnable.
View return policy
Description
CD36 (fatty acid translocase, FAT) is an 88 kDa, ditopic glycosylated protein that belongs to the class B family of scavenger receptors. CD36 is expressed by most resting marginal zone B cells but not by follicular and B1 B cells, and is rapidly induced on Follicular B cells in vitro upon TLR and CD40 stimulation. CD36 does not affect the development of B cells, but modulates both primary and secondary antibody response. Similar to glucose transporter GLUT4, CD36 is translocated from intracellular pools to the plasma membrane following cell stimulation by insulin. In mouse, CD36 is responsible for gustatory perception of long-chain fatty acids. CD36 is preferentially found within lipid rafts, which facilitates its association with receptors, signaling and adapter molecules. CD36 binds to collagen, thrombospondin, anionic phospholipids and oxidized LDL. Further, CD36 may function as a cell adhesion molecule and directly mediates cyto-adherence of Plasmodium falciparum parasitized erythrocytes. Mutations in the CD36 gene cause platelet glycoprotein deficiency. Multiple alternatively spliced transcript variants encoding the CD36 protein have been found.Specifications
| CD36 | |
| Polyclonal | |
| Unconjugated | |
| CD36 | |
| CD36, CHDS7, FAT, Fatty acid translocase, Glycoprotein IIIb, GP3B, GP4, GPIIIB, GPIV, PAS 4, PAS IV, PASIV, Platelet collagen receptor, Platelet glycoprotein 4, Platelet glycoprotein IV, SCARB3, Thrombospondin receptor | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100765055, 12491, 948 | |
| -20°C | |
| Liquid |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P16671, Q08857 | |
| CD36 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human, Hamster | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction