missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD36 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen PA116813
This item is not returnable.
View return policy
Description
This antibody shares 95% sequence homology with rat.
CD36 Polyclonal antibody specifically detects CD36 in Bovine, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.
Specifications
| CD36 | |
| Polyclonal | |
| Unconjugated | |
| CD36 | |
| adipocyte membrane protein; BDPLT10; Cd36; CD36 antigen; CD36 antigen (collagen type I receptor, thrombospondin receptor); CD36 molecule; CD36 molecule (thrombospondin receptor); CD36 protein; CHDS7; cluster determinant 36; collagen type I receptor thrombospondin receptor; collagen type I receptor, thrombospondin receptor; FAT; FAT/CD36; fatty acid translocase; fatty acid translocase/CD36; fatty acid transport protein; glycoprotein IIIb; GP3B; GP4; GPIIIB; GPIV; I79_011940; Leukocyte differentiation antigen CD36; PAS IV; PAS4; PAS-4; PAS-4 protein; PASIV; Platelet collagen receptor; Platelet glycoprotein 4; platelet glycoprotein IV; Scarb3; scavenger receptor class B, member 3; SR-B3; Thrombospondin receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100008825, 12491, 281052, 29184, 733702, 948 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| IgG |
| Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P16671, P26201, Q07969, Q08857 | |
| CD36 | |
| Synthetic peptide corressponding to a region of human CD36 between residues 100-200. | |
| 100 μL | |
| Primary | |
| Bovine, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction