Learn More
CD36 Mouse, anti-Human, Unconjugated, Clone: FA6-152, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60084
This item is not returnable.
View return policy
Description
- The FA6-152 antibody reacts with CD36, an ∼88 kDa transmembrane glycoprotein that functions as a scavenger receptor, cell adhesion molecule, and signal transducer on the surface of many types of cells including platelets, monocytes, macrophages, erythrocyte precursors, endothelial and epithelial cells, and some macrophage-derived dendritic cells
- The topology of the protein is thought to comprise a large extracellular loop with the N- and C-termini both inserted through the cell membrane
- CD36 binds multiple ligands such as thrombospondin, collagen, lipoproteins, and long-chain fatty acids, and has evident roles in the phagocytic clearance of apoptotic cells, inhibition of angiogenesis, metabolism of glucose and fatty acids, inflammation, and the pathogenesis of malaria
- The epitope for FA6-152 reportedly resides within an immunodominant domain of CD36 comprising amino acids 155 - 183
Specifications
| CD36 | |
| Monoclonal | |
| Unconjugated | |
| 100 μg | |
| Human, Rat | |
| Monocytes, Other |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| FA6-152 | |
| Mouse | |
| Primary | |
| IgG1 κ |