missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD36 Antibody (Ona Thera Patent Anti-Cd36), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Marca: Novus Biologicals (Bio-Techne) NBP3-28168CL1
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
CD36 Monoclonal antibody specifically detects CD36 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifica
| CD36 | |
| Monoclonal | |
| CoraFluor 1 | |
| BDPLT10, CD36 antigen (collagen type I receptor, thrombospondin receptor), CD36 molecule (thrombospondin receptor), CHDS7, cluster determinant 36, FAT, Fatty acid translocase, Glycoprotein IIIb, GP3B, GP4, GPIIIB, GPIV, leukocyte differentiation antigen CD36, PAS IV, PAS-4 protein, PASIV, platelet glycoprotein 4, platelet glycoprotein IV, SCARB3, scavenger receptor class B, member 3 | |
| CD36 | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Lipid and Metabolism, Stem Cell Markers | |
| 948 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Ona Thera Patent Anti-Cd36 | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto