missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD36 Antibody (D-2712), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-59724AF750
This item is not returnable.
View return policy
Description
CD36 Monoclonal specifically detects CD36 in Mouse, Rat, Human (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CD36/SR-B3 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CD36 | |
| Adenovirus expressing recombinant mouse CD36. [Uniprot: Q08857] | |
| Primary | |
| Mouse, Rat, Human | |
| IgA κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| D-2712 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor | |
| Mouse | |
| 0.1 mL | |
| 948 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction