missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD36 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 82 publications
£248.00 - £503.00
Specifications
| Antigen | CD36/SR-B3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:200-1:400, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200-1:400, Immunohistochemistry-Frozen reported in scientific literature (PMID 24531551) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18719873
|
Novus Biologicals
NB400-144SS |
0.025 mL |
£248.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18797473
|
Novus Biologicals
NB400-144 |
£503.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
CD36 Polyclonal specifically detects CD36 in Human, Mouse, Rat, Porcine, Bovine, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| CD36/SR-B3 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:200-1:400, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200-1:400, Immunohistochemistry-Frozen reported in scientific literature (PMID 24531551) | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Stem Cell Markers | |
| PBS with 0.02% Sodium Azide | |
| CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor | |
| CD36 | |
| IgG | |
| Affinity Purified | |
| Human, bovine, mouse and monkey. Porcine reactivity reported in scientific literature (PMID: 23727393). Immunogen sequence has 95% identity to rat. |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine, Primate, Rabbit | |
| P16671 | |
| 948 | |
| A synthetic peptide mapping to a region of human CD36 between residues 100-200. [UniProt# P16671] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 75 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title