Learn More
Invitrogen™ CD357 (AITR/GITR) Monoclonal Antibody (110416)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA523854
Description
This antibody does not cross-react with recombinant human (rh) 4-1BB, recombinant mouse (rm) 4-1BB, rhCD27, rmCD27, rhCD30, rmCD30, rhCD40, rmCD40, rhDR3, rhDR6, rhEDAR, rmEDAR, rhFas, rmFAS, rmGITR, rhHVEM, rhLymphotoxin R beta, rmLymphotoxin R beta, rhNGF R, rhOPG, rmOPG, rhRANK, rmRANK, rhTAJ, rhTNF RI, rmTNF RI, rhTNF RII, or rmTNF RII. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Tumor necrosis factor receptor superfamily member 18 (TNFRSF18), also called GITR or AITR is a protein that in humans is encoded by the TNFRSF18 gene. It is mapped to 1p36.33. This gene encodes a member of the TNF-receptor superfamily. The encoded receptor has been shown to have increased expression upon T-cell activation, and it is thought to play a key role in dominant immunological self-tolerance maintained by CD25 (+)CD4 (+) regulatory T cells. Knockout studies in mice also suggest the role of this receptor is in the regulation of CD3-driven T-cell activation and programmed cell death. Three alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
Specifications
| CD357 (AITR/GITR) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9Y5U5 | |
| Tnfrsf18 | |
| Mouse myeloma cell line NS0-derived recombinant human GITR/TNFRSF18 Gln26-Glu161 (Thr43Ala). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Neutralization, Western Blot | |
| 110416 | |
| Unconjugated | |
| Tnfrsf18 | |
| Activation-inducible TNFR family receptor; AITR; CD357; GITR; GITR-D; glucocorticoid-induced TNFR-related protein; PRO364; TNF receptor superfamily activation-inducible protein; TNF receptor superfamily member 18; TNFRSF18; Tumor necrosis factor receptor superfamily member 18; tumor necrosis factor receptor superfamily, member 18; UNQ319; UNQ319/PRO364 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 8784 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.