Learn More
CD355 (CRTAM) Monoclonal Antibody (Cr24.1), PE, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£140.00 - £331.00
Specifications
| Antigen | CD355 (CRTAM) |
|---|---|
| Clone | Cr24.1 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17133873
|
eBioscience
12-3559-41 |
25 Tests | |||||||
17143873
![]() |
Invitrogen™
12-3559-42 |
100 Tests |
£331.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: This monoclonal antibody Cr24.1 reacts with human CD355, also known as Class-I MHC-restricted T-cell-associated molecule (CRTAM). CD355 usually exists as a homodimer, however, it forms a heterodimer with its ligand Necl-2. CD355 is transiently expressed by activated NK, NKT, gamma delta T, and CD8+ T cells. CD355 is also expressed by Purkinje neurons. Human CD355 is involved in immunosurveillance of cancer cells, gamma delta T cell survival, cell polarity, retention of lymphocytes in lymph nodes, promotion of cytotoxicity in NK cells, and IFN gamma secretion from CD8+ T cells. Cr24.1 has been reported to have antagonist activity. Applications Reported: This CR24.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CR24.1 antibody has been pre-diluted and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
The CRTAM gene is upregulated in CD4-positive and CD8-positive T cells and encodes a type I transmembrane protein with V and C1-like Ig domains.Specifications
| CD355 (CRTAM) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| O95727 | |
| CRTAM | |
| Primary | |
| CRTAM |
| Cr24.1 | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| Human | |
| 56253 | |
| IgG2a κ | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.