missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD35 Antibody (S06-6D1), Novus Biologicals™
Rabbit Monoclonal Antibody
£328.00 - £500.00
Specifications
| Antigen | CD35 |
|---|---|
| Clone | S06-6D1 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18615864
|
Novus Biologicals
NBP3-19598-50ul |
50 μg |
£328.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18639064
|
Novus Biologicals
NBP3-19598-100ul |
100 μg |
£500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD35 Monoclonal antibody specifically detects CD35 in Human samples. It is validated for Western BlotSpecifications
| CD35 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| complement component (3b/4b) receptor 1 (Knops blood group), KN | |
| A synthetic peptide of human CD35 (Uniprot # P17927) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S06-6D1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 1378 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title