missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD35 Antibody (HB-8592) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-09019-0.025mg
This item is not returnable.
View return policy
Description
CD35 Monoclonal antibody specifically detects CD35 in Human samples. It is validated for Flow Cytometry
Specifications
| CD35 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| HB-8592 | |
| Unconjugated | |
| complement component (3b/4b) receptor 1 (Knops blood group), KN | |
| Animals were immunized with purified CR1 from human erythrocytes. Spleen cells were fused with P3X63Ag8.653 myeloma cells. | |
| 0.025 mg | |
| Immunology | |
| 1378 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction