missing translation for 'onlineSavingsMsg'
Learn More

CD35 Antibody (8C12), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18329180 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329180 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18329180 Supplier Novus Biologicals Supplier No. NBP281049AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

CD35 Monoclonal specifically detects CD35 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD35
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 8C12
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias complement component (3b/4b) receptor 1 (Knops blood group), KN
Gene Symbols CR1
Host Species Rat
Immunogen Purified murine CR1 protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1378
Test Specificity Recognises an epitope on CD35 (CR1, 190 kDa), but not on CD21 (CR2, 145-150 kDa), which are alternative splice variants of primary Cr2 gene transcripts.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.