missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD340 (HER-2) Mouse anti-Human, BV421, Clone: 24D2, BD Horizon™
Mouse Monoclonal Antibody
£277.98 - £599.20
Specifications
| Antigen | CD340 (HER-2) |
|---|---|
| Clone | 24D2 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15865069
|
BD Biosciences
566459 |
25 μg |
£290.00 £277.98 / Each Save £12.02 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15855069
|
BD Biosciences
566458 |
0.1 mg |
£685.00 £599.20 / Each Save £85.80 12% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD340 (HER-2) Monoclonal antibody specifically detects CD340 (HER-2) in Human samples. It is validated for Flow Cytometry.Specifications
| CD340 (HER-2) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG1 κ | |
| Refrigerate (2 to 8°C) |
| 24D2 | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| Mouse | |
| Human | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title