missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34 Mouse, anti-Human, PE, Clone: 563, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60119PE
This item is not returnable.
View return policy
Description
- CD34 is an ∼105 - 120 kDa type 1 transmembrane glycoprotein expressed on the surface of most human hematopoietic stem and progenitor cells (HSPCs), as well as on endothelial cells and some tumor cell lines
- The frequency of CD34+ cells is low in bone marrow (1 - 4%), cord blood (∼1%), and peripheral blood (< 0.1%)
- CD34 is used to identify and isolate HSPCs, including those capable of reconstituting hematopoiesis after transplantation
- Distinct epitope groups have been assigned to CD34 based on differences in the structure of carbohydrate groups, which affects their sensitivity to enzymatic cleavage
- The 563 antibody recognizes a class III epitope (resistant to neuraminidase and O-glycoprotease)
- Clone 563 also cross-reacts with CD34+ cells of rhesus, cynomolgus, and pigtailed macaques, but not baboons
Specifications
| CD34 | |
| Monoclonal | |
| PE | |
| 50 Tests | |
| Cynomolgus, Human, Rhesus Monkey | |
| Hematopoietic Stem and Progenitor Cells |
| Flow Cytometry | |
| 563 | |
| Mouse | |
| Primary | |
| IgG1 κ |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction