missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34 Mouse, anti-Human, APC, Clone: 581, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60013AZ
This item is not returnable.
View return policy
Description
- The 581 antibody reacts with human CD34, an ∼105 - 120 kDa type 1 transmembrane glycoprotein expressed on the surface of most human hematopoietic stem and progenitor cells (HSPCs) as well as on mesenchymal stem cells, embryonic fibroblasts, endothelial cells, neurons, and some tumor cell lines
- CD34 is expressed only transiently during hematopoiesis, so the frequency of CD34+ cells is low in bone marrow or cord blood (∼1 - 5%) and very low (∼0.1 - 0.5%) in peripheral blood
- CD34 is a marker used to identify and isolate HSPCs capable of cell engraftment
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ Human CD34 Positive Selection Kit II (Catalog #17856) and for labeling human mesenchymal cells grown in MesenCult™ Proliferation Kit (Human; Catalog #05411)
Specifications
| CD34 | |
| Monoclonal | |
| APC | |
| 100 Tests | |
| Human | |
| Hematopoietic Stem and Progenitor Cells |
| CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
| 581 | |
| Mouse | |
| Primary | |
| IgG1 κ |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction