missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34 Antibody (1H6), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3346PECY7
This item is not returnable.
View return policy
Description
CD34 Monoclonal antibody specifically detects CD34 in Canine samples. It is validated for Flow Cytometry
Specifications
| CD34 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Canine | |
| Purified |
| Flow Cytometry | |
| 1H6 | |
| Flow Cytometry | |
| CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule | |
| COS-7 African green monkey SV40 transformed kidney fibroblast-like cell line-derived recombinant canine CD34 | |
| 0.1 mL | |
| Adaptive Immunity, Angiogenesis, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors | |
| 947 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction