Learn More
CD339 (Jagged 1) Monoclonal Antibody (HMJ1-29), Functional Grade, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
£2595.00 - £12950.00
Specifications
| Antigen | CD339 (Jagged 1) |
|---|---|
| Clone | HMJ1-29 |
| Concentration | 1 mg/mL |
| Applications | Functional Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17101232
|
Affymetrix eBioscience
16-3391-38 |
5 mg | |||||||
|
17181222
|
Invitrogen™
16-3391-025 |
25 mg |
£8590.00
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17191222
|
Affymetrix eBioscience
16-3391-050 |
50 mg | |||||||
Descripción
Description: This HMJ1-29 monoclonal antibody reacts with mouse Jagged-1, one of five type I transmembrane proteins that serves as a Notch receptor ligand. Upon binding the Notch receptor (e.g. Notch 1-4), Jagged-1 undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases an intracellular fragment whose biological function remains controversial. Jagged-1 is expressed by thymic lymphoid and stromal cells, as well as macrophages and CD8+CD11c+ dendritic cells in the spleen. The HMJ1-29 antibody is reported to crossreact to rat. Applications Reported: This HMD1-5 antibody has been reported for use in functional assays. Applications Tested: This HMJ1-29 antibody has been tested by flow cytometric analysis of Jagged-1-transfected CHO cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
The jagged 1 protein encoded by JAG1 is the human homolog of the Drosophilia jagged protein. Human jagged 1 is the ligand for the receptor notch 1, the latter a human homolog of the Drosophilia jagged receptor notch. Mutations that alter the jagged 1 protein cause Alagille syndrome. Jagged 1 signalling through notch 1 has also been shown to play a role in hematopoiesis.Especificaciones
| CD339 (Jagged 1) | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| ABE2; AGS; AHD; AWS; CD339; Gsfabe2; HJ1; Htu; JAG1; jagged 1; jagged canonical Notch ligand 1; Jagged1; JAGL1; Ozz; protein jagged-1; Ser-1; Serrate-1 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| HMJ1-29 | |
| Functional Assay | |
| Functional Grade | |
| Armenian Hamster | |
| Mouse, Rat | |
| Q63722, Q9QXX0 | |
| 16449, 29146 | |
| Primary | |
| 4° C | |
| JAG1 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.