missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD338 (ABCG2) Mouse anti-Human, Unconjugated, Clone: 5D3/CD338, BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 552823
This item is not returnable.
View return policy
Description
CD338 (ABCG2) Monoclonal antibody specifically detects CD338 (ABCG2) in Human samples. It is validated for Flow Cytometry.
Specifications
| CD338 (ABCG2) | |
| Monoclonal | |
| 0.5 mg/mL | |
| Mouse | |
| RUO | |
| CD338 (ABCG2) | |
| Refrigerate (2 to 8°C) | |
| IgG2b κ |
| Flow Cytometry | |
| 5D3/CD338 | |
| Unconjugated | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction