missing translation for 'onlineSavingsMsg'
Learn More

CD336 (NKp44) Mouse anti-Human, Brilliant Violet 421, Clone: p44-8, BD Optibuild™

Product Code. 15848929
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15848929 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15848929 Supplier BD Biosciences Supplier No. 744299

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The p44-8 monoclonal antibody specifically binds to the natural killer (NK) cell receptor, NKp44, which is also known as CD336, Natural cytotoxicity triggering receptor 2 (NCR2), or Lymphocyte antigen 95 homolog (Ly95). NKp44 is a ñ44 kDa type I transmembrane protein that belongs to the natural cytotoxicity receptor (NCR) family within the immunoglobulin superfamily. NKp44 is expressed by activated NK cells. NKp44 serves as an activating receptor that can enhance NK cell mediated lysis of target cells including tumor cells and virus-infected cells. Killer activating receptor associated protein (KARAP), which is also known as DAP12, is an intracellular adaptor protein that can associate with the intracellular region of NKp44. DAP12 can then function to help transmit activating signals through its immunoreceptor tyrosine-based activation motif (ITAM).

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD336 (NKp44)
Classification Monoclonal
Clone p44-8
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD336, NCR2, NCTR2, NKp44, NK-p44, LY95, dJ149M18.1
Host Species Mouse
Immunogen Human NKp44
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.