missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD336 (NKp44) Monoclonal Antibody (44.189), PE, eBioscience™
GREENER_CHOICE

Mouse Monoclonal Antibody

£233.00

Specifications

Antigen CD336 (NKp44)
Clone 44.189
Concentration 5 μL/Test
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Applications Flow Cytometry
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15586966
GREENER_CHOICE
View Documents
Invitrogen™
12-3369-42
100 Tests
£233.00
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: The monoclonal antibody 44.189 reacts with human CD336, which is also known as NKp44 or NCR2. CD336 is a 44-kDa transmembrane glycoprotein belonging to the natural cytotoxicity receptor (NCR) family and contains an ITIM domain located in the cytoplasmic tail. Expression of CD336 and NK cell activation by the receptor requires association with DAP12, an ITAM-containing transmembrane accessory protein. Once thought to be expressed exclusively on IL-2-activated NK cells and a subset of gamma delta T lymphocytes, studies have shown expression of CD336 on IL-3-stimulated plasmacytoid dendritic cells, alpha beta T cells in the small intestinal epithelium, and T cells in cord blood. CD336 has been shown to be involved in the innate response of activated NK cells against viral infection and tumor cell proliferation. The 44.189 antibody has been reported to facilitate CD336 crosslinking. Applications Reported: This 44.189 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 44.189 antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Gre...

CD336 is also known as NKp44 or NCR2. CD336 is a 44-kDa transmembrane glycoprotein belonging to the natural cytotoxicity receptor (NCR) family and contains an ITIM domain located in the cytoplasmic tail. Expression of CD336 and NK cell activation by the receptor requires association with DAP12, an ITAM-containing transmembrane accessory protein. Once thought to be expressed exclusively on IL-2-activated NK cells and a subset of gamma delta T lymphocytes, studies have shown expression of CD336 on IL-3-stimulated plasmacytoid dendritic cells, alpha beta T cells in the small intestinal epithelium, and T cells in cord blood. CD336 has been shown to be involved in the innate response of activated NK cells against viral infection and tumor cell proliferation.
Specifications

Specifications

CD336 (NKp44)
5 μL/Test
Flow Cytometry
PE
Mouse
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
O95944
9436
Primary
Affinity chromatography
44.189
4°C, store in dark, DO NOT FREEZE!
Monoclonal
Liquid
IgG2b κ
Human
NCR2
CD336; CD336 antigen; dJ149M18.1; LY95; lymphocyte antigen 95 (activating NK-receptor; NK-p44); Lymphocyte antigen 95 homolog; lymphocyte antigen 95 homolog (activating NK-receptor; NK-p44); natural cytotoxicity triggering receptor 2; Natural killer cell p44-related protein; natural killer cell receptor; NCR2; NK cell activating receptor (NKp44); NK cell-activating receptor; NKP44; NK-p44; NK-p44dJ149M18.1
NCR2
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.