Learn More
CD335 (NKp46) Mouse anti-Human, BB515, Clone: 9E2/NKp46 (also known as 9-E2), BD
Mouse Monoclonal Antibody
Brand:  BD Biosciences 564536
Product Code. 15843709
  • ONLINE EXCLUSIVE PRICE
    £259.00
    £248.58 / Each
    Save £10.42
    4% Off
Description
The 9E2/Nkp46 monoclonal antibody specifically binds to CD335. CD335 is also known as the Natural killer cell p46-related protein (NKp46) and the Natural cytotoxicity triggering receptor 1 (NCR1). CD335 is a 46 kDa type I membrane glycoprotein that is expressed on resting and activated NK cells. Its extracellular region contains two C2-type, Ig-like domains. The transmembrane domain contains a positively charged amino acid (Arg) which could be involved in stabilizing the association with CD3ζ. Its intracellular region does not contain immunoreceptor tyrosine-based activating motifs (ITAM), but it is linked to intracytoplasmic transduction machinery by its association with CD3ζ and FcεRIγ adaptor proteins.CD335 along with NKp30 and NKp44 are referred to as natural cytotoxicity receptors (NCR). These receptors play very important roles in cells that kill virus-infected target cells, tumor cells and MHC-class I-unprotected cells.
The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490 nm and an emission max of 515 nm, BD Horizon BB515 can be excited by the 488 nm laser and detected in a standard FITC set (eg, 530/30nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector. BB515 is a dye that was exclusively developed by BD Biosciences as a brighter alternative to FITC. This dye is up to seven times brighter than FITC and has less spillover into the PE channel. Due to similar excitation and emission properties, BB515, FITC, and Alexa Fluor™ 488 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD335
9E2/NKp46 (also known as 9-E2)
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
Affinity Purified
RUO
Human
IgG1 κ
Monoclonal
Brilliant Blue 515 (BB515)
NCR1, NK-p46, hNKp46, LY94, Natural cytotoxicity triggering receptor 1
Human NKp46 Recombinant Protein
100 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.