missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD33 Mouse anti-Human, BV421, Clone: WM53, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 565949
This item is not returnable.
View return policy
Description
CD33 Monoclonal antibody specifically detects CD33 in Human samples. It is validated for Flow cytometry.
Specifications
| CD33 | |
| Monoclonal | |
| Brilliant Violet 421 | |
| Siglec-3; SIGLEC3; Sialic acid-binding Ig-like lectin 3; p67; gp67; My9 | |
| Acute Myeloid Leukemia Blasts | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. | |
| IgG1 κ |
| Flow Cytometry | |
| WM53 (also known as WM-53) | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 25 Tests | |
| Cell Biology | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction