Learn More
CD33 Monoclonal Antibody (WM-53 (WM53)), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-0338-95
Description
Description: The WM-53 monoclonal antibody reacts with human CD33, also known as GP67 and P67, a 67 kDa type I transmembrane glycoprotein that is a member of the Siglec (sialic acid-binding Ig superfamily lectin) family. It is highly specific to the hematopoietic compartment and is expressed on monocytes, activated T cells, granulocytes, myeloid progenitors, and mast cells. Applications Reported: The WM-53 (WM53) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunocytochemistry staining. Applications Tested: The WM-53 (WM53) antibody has been tested by flow cytometric analysis of lysed whole blood. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
CD33 Monoclonal antibody specifically detects CD33 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Specifications
| CD33 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P20138 | |
| Mouse | |
| 10 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| WM-53 (WM53) | |
| Unconjugated | |
| CD33 | |
| Cd33; CD33 antigen; CD33 antigen (gp67); CD33 molecule; CD33 molecule transcript; FLJ00391; gp67; Myeloid cell surface antigen CD33; p67; sialic acid binding Ig-like lectin 3; sialic acid-binding Ig-like lectin 3; SIGLEC3; SIGLEC-3 | |
| Affinity chromatography | |
| RUO | |
| 945 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.