Learn More
Invitrogen™ CD328 (Siglec7) Monoclonal Antibody (S7.7)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528200
Description
Mouse anti Human Siglec-7 antibody, cloneS7.7 recognizes human Sialic acid-binding Ig-like lectin 7, otherwise known as CD328, a putative adhesion molecule and member of the Ig superfamily, predominantly expressed by both resting and activated natural killer (NK) cells, and also at lower levels by monocytes, granulocytes, monocyte-derived macrophages and dendritic cells, and a small population of CD8+ memory T cells.
Putative adhesion molecule that mediates sialic-acid dependent binding to cells. Preferentially binds to alpha-2,3- and alpha-2,6-linked sialic acid. Also binds disialogangliosides (disialogalactosyl globoside, disialyl lactotetraosylceramide and disialyl GalNAc lactotetraoslylceramide). The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface. In the immune response, may act as an inhibitory receptor upon ligand induced tyrosine phosphorylation by recruiting cytoplasmic phosphatase(s) via their SH2 domain(s) that block signal transduction through dephosphorylation of signaling molecules. Mediates inhibition of natural killer cells cytotoxicity. May play a role in hemopoiesis. Inhibits differentiation of CD34+ cell precursors towards myelomonocytic cell lineage and proliferation of leukemic myeloid cells (in vitro).
Specifications
| CD328 (Siglec7) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9Y286 | |
| SIGLEC7 | |
| Siglec-7 3T3 transfected cells. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot | |
| S7.7 | |
| Unconjugated | |
| SIGLEC7 | |
| Adhesion inhibitory receptor molecule 1; adhesion inhibitory receptor molecule 1, siglec-7; AIRM1; AIRM-1; CD328; CDw328; D-siglec; p75; p75/ AIRM-1; p75/AIRM1; QA79; QA79 membrane protein; sialic acid binding Ig like lectin 7; sialic acid binding Ig-like lectin 7; sialic acid binding immunoglobulin-like lectin 7; sialic acid-binding Ig-like lectin 7; Siglec; SIGLEC19P; SIGLEC7; SIGLEC-7; SIGLECP2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 27036 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.