missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD324 (E-Cadherin) Rat anti-Mouse, Human, PE, Clone: DECMA-1, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 567052
This item is not returnable.
View return policy
Description
CD324 (E-Cadherin) Monoclonal antibody specifically detects CD324 (E-Cadherin) in Mouse, Human samples. It is validated for Flow Cytometry
Specifications
| CD324 (E-Cadherin) | |
| Monoclonal | |
| PE | |
| ARC-1; Cdh1; E-cad; Ecad; UVO; cadherin-1; epithelial cadherin; uvomorulin | |
| Mouse teratocarcinoma Cell Line | |
| RUO | |
| Mouse, Human | |
| IgG1 κ |
| Flow Cytometry | |
| DECMA-1 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction