missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD323 (JAM3) Monoclonal Antibody (CRAM-18 F26)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA528231
This item is not returnable.
View return policy
Description
Rat anti Mouse JAM-c antibody, clone CRAM-18 F26 recognizes mouse and human Junctional adhesion molecule C (JAM-C), also known as JAM-3 and, historically, as JAM-2.
JAM3 is involved in tight junctions and represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. The protein encoded by this immunoglobulin superfamily gene member is localized in the tight junctions between high endothelial cells. Unlike other proteins in this family, the this protein is unable to adhere to leukocyte cell lines and only forms weak homotypic interactions. The encoded protein is a member of the junctional adhesion molecule protein family and acts as a receptor for another member of this family.
Specifications
| CD323 (JAM3) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9D8B7 | |
| JAM3 | |
| Recombinant soluble JAM-C. | |
| 1 mg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunocytochemistry | |
| CRAM-18 F26 | |
| Unconjugated | |
| JAM3 | |
| 1110002N23Rik; JAM-2; JAM3; JAM-3; JAMC; JAM-C; Jcam3; junction adhesion molecule 3; junction cell adhesion molecule 3; junction cell adhesion molecule C; junctional adhesion molecule 3; Junctional adhesion molecule C; sJAM-C; Soluble form of JAM-C; UNQ859/PRO1868 | |
| Rat | |
| Protein G | |
| RUO | |
| 83964 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction