missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD320 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581338
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Mouse CD320/8D6 antigen.
Germinal center-B (GC-B) cells differentiate into memory B-cells and plasma cells (PC) through interaction with T-cells and follicular dendritic cells (FDC). CD320 augments the proliferation of PC precursors generated by IL-10. Receptor for the cellular uptake of transcobalamin bound cobalamin.
Specifications
| CD320 | |
| Polyclonal | |
| Unconjugated | |
| Cd320 | |
| 425O18-1; 8D6; 8D6 antigen; 8D6A; 8D6AFDC-SM-8D6; 8D6TCBLR; Cd320; CD320 antigen; CD320 molecule; D17Ertd716e; FDC-signaling molecule 8D6; FDC-SM-8D6; NG29; putative VLDL lipoprotein receptor; RGD1305860; TCblR; transcobalamin receptor; UNQ198/PRO224; VLDL | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 54219 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9Z1P5 | |
| Cd320 | |
| Recombinant Mouse CD320/8D6 antigen Protein (Met1-Gly208). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction