missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD32 Rat, PE, Clone: 2.4G2, enQuireBio™
Rat Monoclonal Antibody
£81.75 - £243.00
Specifications
Antigen | CD32 |
---|---|
Clone | 2.4G2 |
Applications | Flow Cytometry |
Conjugate | PE |
Host Species | Rat |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15972588
|
EnquireBio
QAB87-PE-25UG |
25 μg |
£81.75
25µg |
Estimated Shipment: 21-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15962588
|
EnquireBio
QAB87-PE-100UG |
100 μg |
£243.00
100µg |
Estimated Shipment: 21-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Specifications
CD32 | |
Flow Cytometry | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
14131 | |
Protein A/G Purified |
2.4G2 | |
PE | |
RUO | |
XP_006496722 | |
IgG2b | |
Store at 2-4°C. |