Learn More
CD32 Mouse, anti-Human, Unconjugated, Clone: FLI8.26, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60135
This item is not returnable.
View return policy
Description
- The FLI8.26 antibody reacts with human CD32 (FcγRII), an ∼40 kDa type 1 transmembrane glycoprotein that mediates several functions including phagocytosis, cytotoxicity, immunomodulation, and platelet aggregation
- The FLI8.26 antibody cross-reacts with monocytes, granulocytes, and platelets
- CD32 is encoded by three genes (A, B, C) and at least 6 isoforms are generated via alternative mRNA splicing, i.e., IIa1, IIa2, IIb1, IIb2, IIb3 and IIc
- Clone FLI8.26 inhibits binding of the FcγRIIa-specific clone IV.3 antibody in flow cytometry analyses
- CD32 binds weakly to the Fc region of monomeric IgG but more strongly to IgG aggregates and immune complexes
- These interactions can result in non-specific labeling in antibody-based detection and cell separation experiments and the FLI8.26 antibody may be used as a blocking antibody to reduce non-specific binding
Specifications
| CD32 | |
| Monoclonal | |
| Unconjugated | |
| 100 μg | |
| Baboon, Cynomolgus, Rhesus Monkey, Chimpanzee, Human | |
| B Cells, Granulocytes and Subsets, Monocytes |
| Flow Cytometry | |
| FLI8.26 | |
| Mouse | |
| Primary | |
| IgG2b κ |