missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD32 Mouse anti-Human, BUV737, Clone: FLI8.26 (also known as 8.26), BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 741835
This item is not returnable.
View return policy
Description
CD32 Monoclonal antibody specifically detects CD32 in Human samples. It is validated for Flow Cytometry
Specifications
| CD32 | |
| Monoclonal | |
| Brilliant Ultraviolet 737 | |
| CD32a,FcγRIIa, FCGR2A; CD32b, FcγRIIb, FCGR2B; CD32c, FcγRIIc, FCGR2C | |
| K562 or FcγRII+ L cell lines | |
| RUO | |
| Human | |
| IgG2b κ |
| Flow Cytometry | |
| FLI8.26 (also known as 8.26) | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV737 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction