Learn More
CD32 Mouse anti-Human, BUV395, Clone: 3D3, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 744259
Description
The 3D3 monoclonal antibody specifically recognizes FcγRII (CD32), a 40 kDa, polymorphic type I transmembrane glycoprotein that serves as a low affinity receptor for aggregated IgG. This highly glycosylated molecule (encoded by at least two different genes) is expressed on monocytes, granulocytes, platelets and B cells. Unlike the FLI28.26 mAb, the 3D3 mAb detected a polymorphic CD32 antigen expressed on B cells of all donors, but only on platelets, monocytes and granulocytes of some donors. The platelets from 3D3+ donors respond to certain stimulatory mAb such as CD165 (clone SN2) which results in aggregation. On the other hand, the platelets from 3D3 negative donors do not form aggregates after stimulation. Individuals can be divided into two groups as responder and non-responder depending on expression, or non-expression, of 3D3. In comparison to the 3D3 mAb, the FLI8.26 mAb detects a monomorphic CD32 antigen expressed on all human donors.
The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
Specifications
| CD32 | |
| 3D3 | |
| Brilliant Ultraviolet 395 | |
| FcγRII, CD32a/FcγRIIa/FCGR2A, CD32b/FcγRIIb/FCGR2B | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.