Learn More
CD32 Monoclonal Antibody (6C4 (CD32)), Biotin, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£147.00 - £368.00
Specifications
| Antigen | CD32 |
|---|---|
| Clone | 6C4 (CD32) |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
Description: This 6C4 monoclonal antibody reacts with human CD32 (also known as Fc gammaRII). This antibody recognizes two isoforms of the receptor, Fc gammaRIIA and Fc gammaRIIB. These 40-kDa polymorphic transmembrane glycoproteins are expressed on B cells, granulocytes, monocytes, macrophages, and platelets. Moreover, these receptors are detected on natural killer cells. CD32 enables interaction between Fc gammaRII-expressing cells and opsonized antigen or IgG-containing immune complexes. Both receptors exhibit low affinity towards IgG and play a role in inflammation and autoimmune disease. This clone has been reported to inhibit Ig binding in a rosette blocking assay. Applications Reported: This 6C4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 6C4 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.Specifications
| CD32 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P12318, P31994, P31995 | |
| FCGR2A, FCGR2B, FCGR2C | |
| Primary | |
| Antibody |
| 6C4 (CD32) | |
| Flow Cytometry | |
| Biotin | |
| Mouse | |
| Human | |
| 2212, 2213, 9103 | |
| IgG1 κ | |
| Affinity chromatography |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.