missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD32 (FcγRII) Mouse anti-Human, BV750, Clone: FLI8.26 (also known as 8.26), BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 747477
This item is not returnable.
View return policy
Description
CD32 (FcγRII) Monoclonal antibody specifically detects CD32 (FcγRII) in Human samples. It is validated for Flow Cytometry
Specifications
| CD32 (FcγRII) | |
| Monoclonal | |
| Brilliant Violet 750 | |
| CD32a,FcγRIIa, FCGR2A; CD32b, FcγRIIb, FCGR2B; CD32c, FcγRIIc, FCGR2C | |
| K562 or FcγRII+ L cell lines | |
| RUO | |
| Human | |
| IgG2b κ |
| Flow Cytometry | |
| FLI8.26 (also known as 8.26) | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV750 under optimal conditions that minimize unconjugated dye and antibody. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur