Learn More
CD319 (CRACC) Rat anti-Mouse, Brilliant Violet 786, Clone: 4G2, BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 747993
This item is not returnable.
View return policy
Description
CD319 (CRACC) Monoclonal antibody specifically detects CD319 (CRACC) in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD319 (CRACC) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Rat | |
| RUO | |
| CD319 (CRACC) | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| 4G2 | |
| Brilliant Violet 786 | |
| 50 μg | |
| Primary | |
| Mouse | |
| IgG1 κ |