Learn More
CD317 (BST2, PDCA-1) Monoclonal Antibody (eBio129c (129c)), PE, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£81.00 - £252.00
Specifications
Antigen | CD317 (BST2, PDCA-1) |
---|---|
Clone | eBio129c (129c) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15288739
|
Affymetrix eBioscience
12-3171-82 |
100 μg |
£252.00
100µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15278739
|
Affymetrix eBioscience
12-3171-81 |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The eBio129c monoclonal antibody reacts with mouse PDCA-1 (BST2, CD317), a specific marker of plasmacytoid dendritic cells (pDC), also known as type I IFN-producing cells (IPC) in the naive mouse. Mouse IPCs are typically CD11c+, CD11b-, B220+, Ly-6C+, and CD62L+. PDCA-1 is predomitly expressed by IPCs in the naive mouse which represent sa very minor population (<0.5%) of splenocytes. It is upregulated on numerous cell types following stimulation which triggers an IFN response. PDCA-1 cycles between cell surface and intracellular compartments and may function to regulate trafficking of secreted cytokines. PDCA-1 (BST2) is the protein recognized by the antibody 120G8. The epitope recognized by eBio129c is distinct from eBio927; thus, the antibodies can be used to costain, purify and identify pDCs. Applications Reported: This eBio129c (129c) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio129c (129c) antibody has been tested by flow cytometric analysis of SJL splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
BST2 (CD317, HM1.24 antigen, DAMP-2, tetherin) is an integral membrane protein that is involved in the development and growth of B-cells. The surface expression of BST2 on fibroblast cell lines facilitated the stromal cell-dependent growth of DW34, a pre-B-cell line. BST2 is highly expressed during B-cell development, from pro-B precursors to plasma cells, T-cells, monocytes, NK cells and DCs (at protein level), and myeloma cells. Research studies have shown that BST2 also functions as an inhibitor of retrovirus release from human cells whose activity is antagonized by the HIV-1 accessory protein, Vpu. While BST2 causes retention of virions on cell surfaces or endocytosis into BST2-positive compartments, its depletion abolished the viral requirement for Vpu for virus release, an activity that may represent a potential therapeutic strategy for the treatment of HIV/AIDS. Another disease associated with BST2 dysfunction is stomatitis.Specifications
CD317 (BST2, PDCA-1) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Q8R2Q8 | |
Bst2 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Bst2 |
eBio129c (129c) | |
Flow Cytometry | |
PE | |
Rat | |
Mouse | |
69550 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.