Learn More
CD314 (NKG2D) Monoclonal Antibody (A10), Functional Grade, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
£94.50 - £386.00
Specifications
Antigen | CD314 (NKG2D) |
---|---|
Clone | A10 |
Concentration | 1 mg/mL |
Applications | Flow Cytometry, Functional Assay |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15288657
|
Affymetrix eBioscience
16-5872-82 |
100 μg |
£150.00
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15298657
|
Affymetrix eBioscience
16-5872-85 |
500 μg |
£237.00
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15278657
|
Affymetrix eBioscience
16-5872-81 |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15208667
|
Affymetrix eBioscience
16-5872-86 |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The A10 monoclonal antibody reacts with the mouse NKG2D, a lectin-like molecule expressed on both human and mouse NK cells. Mouse NKG2D binds to retinoic acid-inducible RAE-1 alpha, beta, gamma, delta, epsilon and the minor histocompatibility molecule H60 and has the ability to costimulate multiple NK activation receptors, through the DAP12/DAP10 adaptor molecules. NKG2D is expressed by all spleen and liver NK cells, NK1.1(+) thymocytes, in vitro activated LAK cells, and a subset of splenic NKT cells. A10 and C7 antibodies detect NK cells from all inbred strains of mice tested so far. In redirected lysis assays, A10 is reported to stimulate NK cells to kill Daudi targets. In addition, immobilized A10 is reported to induce production of GM-CSF by NK clones and LAK cells through a CD16 (FC gamma RIII) concomitant engagement. A10 and another hamster anti-mouse NKG2D (clone C7) compete with each other for binding to transfected cells by flow cytometric analysis, suggesting that they may bind to similar epitopes or block each other by steric hindrance. C7 (neutralizing) and A10 (activating) also exhibit different functional properties. Expression of the NKG2D antigen on mouse peripheral NK and NKT cells can be detected by flow cytometric analysis using mAb CX5 with much brighter intensity. Applications Reported: The A10 antibody has been reported for use in flow cytometric analysis. It has also been reported in activation in functional studies.
Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. NK cells can regulate specific humoral and cell-mediated immunity, and preferentially express several calcium-dependent (C-type) lectins, which have been implicated in the regulation of NK cell function. The NK gene encodes a member of the NKG2 family, and the encoded transmembrane protein is characterized by a type II membrane orientation (extracellular C terminus) and the presence of a C-type lectin domain. The NKG2 gene family is located within the NK complex, a region that contains several C-type lectin genes preferentially expressed in NK cells.Specifications
CD314 (NKG2D) | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.2 | |
CD314; D12S2489E; D6H12S2489E; killer cell lectin like receptor K1; Killer cell lectin-like receptor subfamily K member 1; killer cell lectin-like receptor subfamily K, member 1; KLR; Klrk1; natural killer cell group 2D; natural killer cell receptor; NK cell receptor D; NK lectin-like receptor; Nkg2d; NKG2-D; NKG2-D type II integral membrane protein; NKG2-D-activating NK receptor; NKLLR; Nkrp2; NKR-P2; NKR-P2, ortholog of human NKG2D | |
Klrk1 | |
Primary | |
4° C | |
Klrk1 |
A10 | |
Flow Cytometry, Functional Assay | |
Functional Grade | |
Armenian Hamster | |
Mouse | |
O54709 | |
27007 | |
IgG | |
Affinity chromatography | |
Antibody |
For Research Use Only.