Learn More
CD314 (NKG2D) Monoclonal Antibody (1D11), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£90.00 - £233.00
Specifications
Antigen | CD314 (NKG2D) |
---|---|
Clone | 1D11 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE-Cyanine7 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15558586
|
Affymetrix eBioscience
25-5878-42 |
100 Tests |
£233.00
100 tests |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15548586
|
Affymetrix eBioscience
25-5878-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. They can also regulate specific humoral and cell-mediated immunity. NK cells preferentially express several calcium-dependent (C-type) lectins, which have been implicated in the regulation of NK cell function. This gene encodes a member of the NKG2 family, and the encoded transmembrane protein is characterized by a type II membrane orientation (extracellular C terminus) and the presence of a C-type lectin domain. The NKG2 gene family is located within the NK complex, a region that contains several C-type lectin genes preferentially expressed in NK cells.
CD314 (NKG2D) Monoclonal antibody specifically detects CD314 (NKG2D) in Human samples. It is validated for Flow Cytometry.Specifications
CD314 (NKG2D) | |
Flow Cytometry | |
PE-Cyanine7 | |
Mouse | |
Human | |
22914 | |
Primary | |
Antibody |
1D11 | |
Monoclonal | |
Liquid | |
RUO | |
P26718 | |
IgG1 κ | |
Affinity chromatography |
For Research Use Only.