Learn More
CD31 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532321
Description
This antibody is predicted to react with mouse samples based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is tonsil tissue.
CD31 is a glycoprotein expressed on endothelial cells and in platelets. It is known to be involved in cell signaling and cell adhesion.
Specifications
| CD31 | |
| Polyclonal | |
| Unconjugated | |
| P16284, Q08481, Q95242 | |
| PECAM1 | |
| Synthetic peptide corresponding to C-terminus of mouse CD31 protein. | |
| 500 μL | |
| Primary | |
| Human, Mouse, Pig | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| 0.018 mg/mL | |
| PECAM1 | |
| CD31 antigen, CD31/EndoCAM, GPIIA′, PECA1, PECAM-1, CD31/EndoCAM, adhesion molecule, endoCAM, platelet endothelial cell adhesion molecule | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 396941, 5175, 18613 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.