Learn More
CD31/PECAM-1, Mouse anti-Bovine,Goat,Sheep, FITC, Clone: CO.3E-1D4, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-65900F-100UL
Product Code. 18206343
  • £491.00 / 100µL
Description
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Bovine, Equine, Goat, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD31/PECAM-1
Monoclonal
1.0 mg/mL
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet/endothelial cell adhesion molecule
Mouse
Protein G purified
RUO
Primary
NB100-65900 is specific for ovine CD31, also known as PECAM-1. Ovine CD31 is predominantly expressed by peripheral blood platelets and a small percentage of lymphocytes. CD31 is also highly expressed by ovine endothelial cells. Clone CO.3E1D4 is reported to inhibit homotypic leucocyte aggregation induced by anti CD43 antibodies (2). Removal of sodium azide is recommended prior to use in fuctional assays.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2a
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
CO.3E-1D4
FITC
PBS
PECAM1
Ovine leucocytes
100 μL
Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers
5175
Bovine, Goat, Sheep
Purified