missing translation for 'onlineSavingsMsg'
Learn More

CD31 (PECAM-1) Monoclonal Antibody (390), Functional Grade, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Brand:  Invitrogen 16-0311-025

 View more versions of this product

Product Code. 17180982

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

The 390 monoclonal antibody reacts with mouse CD31, also known as platelet-endothelial cell adhesion molecule-1 (PECAM-1) and gpIIa. This 130-140 kDa surface protein is expressed by endothelial cells and at low levels on all leukocytes and platelets. It has been reported that CD38 binds to CD31. Homotypic interaction of CD31 is important in adhesion, cell-cell and cell-matrix interaction, and signal transduction. Applications Reported: The 390 antibody has been reported for use in flow cytometric analysis. 390 has also been reported in in vitro functional studies. Applications Tested: The 390 antibody has been tested by flow cytometric analysis of mouse thymocytes and splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/ μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.

CD31 (PECAM-1) Monoclonal antibody specifically detects CD31 (PECAM-1) in Mouse samples. It is validated for Flow Cytometry, Functional Assay
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD31 (PECAM-1)
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.2
Q08481
Rat
25 mg
Primary
Mouse
Antibody
IgG2a κ
Flow Cytometry, Functional Assay
390
Functional Grade
Pecam1
C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; Fc receptor-like 4-like; GPIIA'; I79_008304; LOC100357605; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; Platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule
Affinity chromatography
RUO
18613
4° C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.