missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD31/PECAM-1 Antibody (CL14693), Novus Biologicals™
Mouse Monoclonal Antibody
£246.00 - £386.00
Specifications
| Antigen | CD31/PECAM-1 |
|---|---|
| Clone | CL14693 |
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:5000-1:10000 |
| Applications | Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043206
|
Novus Biologicals™
NBP3-26877-25ul |
25 μL |
£246.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043129
|
Novus Biologicals™
NBP3-26877-100ul |
100 μL |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD31/PECAM-1 Monoclonal antibody specifically detects CD31/PECAM-1 in Human samples. It is validated for ImmunohistochemistrySpecifications
| CD31/PECAM-1 | |
| 1 mg/mL | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Cytoskeleton Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers | |
| PBS, pH 7.2, 40% glycerol | |
| 5175 | |
| IgG1 | |
| Protein A purified |
| CL14693 | |
| Immunohistochemistry 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
| This antibody was developed against a Synthetic Peptide of P16284. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title